missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Benzyl-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63233.06

Code : NEW

Additional Details : CAS Number : 65795-95-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15409828

  • £149.46 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-Benzyl-5-nitroindole
252.27
1252504
65795-95-1
MFCD03206550

Specifications

1-Benzyl-5-nitroindole
Solid
5g
MFCD03206550
65795-95-1
97%
252.27
1252504
Videos
SDS