missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carboxylic acid, ≥95%, Maybridge

Brand:  Maybridge SPB03031DA

Code : 31

Additional Details : CAS Number : 100957-85-5 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10325542

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carboxylic acid
95.0
95%
258.32
100957-85-5
95%
C15H18N2O2
MFCD02090479

Specifications

1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carboxylic acid
95.0
95%
1g
258.32
100957-85-5
Amber Glass Bottle
95%
C15H18N2O2
MFCD02090479
Videos
SDS