missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-piperidone hydrate hydrochloride, 95%, ACROS Organics™

Brand:  Acros Organics 166920050

Code : 29

Additional Details : CAS Number : 50606-58-1 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10515881

  • £98.60 / 5g


Estimated Shipment
13-03-2018
Add to basket


Specifications

Chemical Identifiers

1-Benzyl-3-piperidone hydrate hydrochloride
50606-58-1
100.0
95%
225.72
95%
94.0
≥94% (on free base)
HCl·xH2O
MFCD00150096

Specifications

1-Benzyl-3-piperidone hydrate hydrochloride
50606-58-1
100.0
Glass bottle
≥94% (on free base)
8% max. (K.F.)
225.72
Cream to pale brown
95%
94.0
Authentic
Powder
95%
HCl·xH2O
MFCD00150096
5g
Videos
SDS