missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-piperidinol, 97%, Thermo Scientific™

Catalog Number p-2695580
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
105973-51-1
C12H18ClNO
227.732
MFCD00044200
PHJDPNCYJSWYGY-UHFFFAOYSA-N
1-benzyl-3-piperidinol hydrochloride, 1-benzylpiperidin-3-ol hydrochloride, 1-benzyl-3-piperidinol hcl, 1-benzyl-3-piperidinolhydrochloride, 1-benzyl-3-hydroxypiperidine hydrochloride, acmc-1ca0o, 1-benzylpiperidin-3-ol, chloride, 1-benzyl3-hydroxypiperidine hydrochloride, 1-phenylmethyl-3-piperidinol hydrochloride, 1-phenylmethyl piperidin-3-ol hydrochloride
16218306
1-benzylpiperidin-3-ol;hydrochloride
C1CC(CN(C1)CC2=CC=CC=C2)O.Cl
This item is not returnable. View return policy
105973-51-1
C12H18ClNO
227.732
MFCD00044200
PHJDPNCYJSWYGY-UHFFFAOYSA-N
1-benzyl-3-piperidinol hydrochloride, 1-benzylpiperidin-3-ol hydrochloride, 1-benzyl-3-piperidinol hcl, 1-benzyl-3-piperidinolhydrochloride, 1-benzyl-3-hydroxypiperidine hydrochloride, acmc-1ca0o, 1-benzylpiperidin-3-ol, chloride, 1-benzyl3-hydroxypiperidine hydrochloride, 1-phenylmethyl-3-piperidinol hydrochloride, 1-phenylmethyl piperidin-3-ol hydrochloride
16218306
1-benzylpiperidin-3-ol;hydrochloride
C1CC(CN(C1)CC2=CC=CC=C2)O.Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 227.73
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-3-piperidinol hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'