missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Benzyl-3-(hydroxymethyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H64403.06

Code : NEW

Additional Details : CAS Number : 5731-17-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451599

  • £150.52 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

1-Benzyl-3-(hydroxymethyl)pyrrolidine
191.27
146199
5731-17-9
MFCD04038527
N-Benzyl-DL-β-prolinol

Specifications

1-Benzyl-3-(hydroxymethyl)pyrrolidine
1.5420
Liquid
137° to 145°C (1mmHg)
191.27
146199
5731-17-9
Pale yellow to brown
97%
5g
MFCD04038527
N-Benzyl-DL-β-prolinol
Videos
SDS