missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Benzyl-3-(4-fluorobenzyl)urea, 95%

Brand:  Alfa Aesar™ H59115.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445328

  • £358.49 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

1-Benzyl-3-(4-fluorobenzyl)urea
258.3
MFCD08890332

Specifications

1-Benzyl-3-(4-fluorobenzyl)urea
95%
258.3
Solid
1g
MFCD08890332
Videos
SDS