missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-(chloromethyl)-1H-imidazole hydrochloride, 97%, Maybridge

Brand:  Maybridge KM08397DE

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 19276-03-0 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 10352335

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

1-Benzyl-2-(chloromethyl)-1H-imidazole hydrochloride
97.0
97%
243.14
19276-03-0
97%
C11H11ClN2·ClH
MFCD00035210

Specifications

1-Benzyl-2-(chloromethyl)-1H-imidazole hydrochloride
97.0
97%
C11H11ClN2·ClH
MFCD00035210
19276-03-0
97%
5g
243.14
Documentation

Technical Documents