missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-2695535
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
3541-37-5
C9H6OS
162.21
MFCD01075041
NXSVNPSWARVMAY-UHFFFAOYSA-N
benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
736500
1-benzothiophene-2-carbaldehyde
O=CC1=CC2=CC=CC=C2S1
This item is not returnable. View return policy
3541-37-5
C9H6OS
162.21
MFCD01075041
NXSVNPSWARVMAY-UHFFFAOYSA-N
benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
736500
1-benzothiophene-2-carbaldehyde
O=CC1=CC2=CC=CC=C2S1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 1 g
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'