Learn More
1-Benzofuran-5-carbaldehyde, 97%, Thermo Scientific™
Brand: Thermo Scientific Maybridge CC07104DE
Code : 31
Additional Details : CAS Number : 10035-16-2 Weight : 0.05500kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
10035-16-2 | |
146.145 | |
LLLBDLDNTMMZHL-UHFFFAOYSA-N | |
2773875 | |
C1=CC2=C(C=CO2)C=C1C=O |
C9H6O2 | |
MFCD03411182 | |
benzofuran-5-carbaldehyde, 5-formylbenzofuran, 5-benzofurancarboxaldehyde, benzofuran-5-carboxaldehyde, 5-formyl-1-benzofuran, 5-formylbenzo b furan, benzo b furan-5-carboxaldehyde, 1-benzofuran-5-carboxaldehyde, benzo b furan-5-carbaldehyde | |
1-benzofuran-5-carbaldehyde |
Specifications
1-Benzofuran-5-carbaldehyde | |
96.5 | |
97% | |
MFCD03411182 | |
benzofuran-5-carbaldehyde, 5-formylbenzofuran, 5-benzofurancarboxaldehyde, benzofuran-5-carboxaldehyde, 5-formyl-1-benzofuran, 5-formylbenzo b furan, benzo b furan-5-carboxaldehyde, 1-benzofuran-5-carboxaldehyde, benzo b furan-5-carbaldehyde | |
LLLBDLDNTMMZHL-UHFFFAOYSA-N | |
1-benzofuran-5-carbaldehyde | |
2773875 | |
97% |
10035-16-2 | |
100.0 | |
C9H6O2 | |
Amber Glass Bottle | |
5g | |
C1=CC2=C(C=CO2)C=C1C=O | |
146.145 | |
146.14 |