Learn More
Thermo Scientific Maybridge 1-Benzofuran-3-carboxylic acid, 1g, 97%
Brand: Thermo Scientific Maybridge CC27501DA
1-Benzofuran-3-carboxylic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 26537-68-8 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC27501DA
1-Benzofuran-3-carboxylic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 26537-68-8 Weight : 0.05100kg