missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylazetan-3-ol, Thermo Scientific™

Catalog Number p-2695463
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
18621-17-5
C16H17NO
239.32
MFCD00205109
MMAJXKGUZYDTHV-UHFFFAOYSA-N
1-benzhydrylazetan-3-ol, 1-diphenylmethyl-3-hydroxyazetidine, 1-diphenylmethyl azetidin-3-ol, 1-benzhydryl-3-azetidinol, 1-benzhydryl-3-hydroxyazetidine, 1-benzhydryl-azetidin-3-ol, n-benzhydrylazetidin-3-ol, 3-azetidinol, 1-diphenylmethyl, 1-diphenylmethyl-3-azetidinol
330448
OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
18621-17-5
C16H17NO
239.32
MFCD00205109
MMAJXKGUZYDTHV-UHFFFAOYSA-N
1-benzhydrylazetan-3-ol, 1-diphenylmethyl-3-hydroxyazetidine, 1-diphenylmethyl azetidin-3-ol, 1-benzhydryl-3-azetidinol, 1-benzhydryl-3-hydroxyazetidine, 1-benzhydryl-azetidin-3-ol, n-benzhydrylazetidin-3-ol, 3-azetidinol, 1-diphenylmethyl, 1-diphenylmethyl-3-azetidinol
330448
OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 1-benzhydrylazetidin-3-ol
Formula Weight 239.32
Percent Purity 97%
Chemical Name or Material 1-Benzhydrylazetan-3-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'