missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Benzenesulfonyl-5-cyanoindole, 97%

Brand:  Alfa Aesar™ H63313.06

Code : NEW

Additional Details : CAS Number : 144340-21-6 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15419948

  • £239.56 / 5g


Estimated Shipment date
from Supplier 15-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Benzenesulfonyl-5-cyanoindole
282.32
UN3439
1-(Benzenesulfonyl)indole-5-carbonitrile
144340-21-6
MFCD09965529
7644292

Specifications

1-Benzenesulfonyl-5-cyanoindole
Solid
282.32
UN3439
1-(Benzenesulfonyl)indole-5-carbonitrile
5g
144340-21-6
97%
MFCD09965529
7644292
Off-white to white
Videos
SDS