missing translation for 'onlineSavingsMsg'
Learn More

1-BCP, Tocris Bioscience™

Modulates AMPA-mediated responses

Brand:  Tocris (Bio-Techne) 104850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438383

  • £312.85 / 50mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A centrally active selective potentiator of AMPA-mediated responses. Facilitates memory retention in rats.
Specifications

Chemical Identifiers

34023-62-6
233.267
1-bcp, 1-1,3-benzodioxol-5-ylcarbonyl piperidine, piperidine, 1-1,3-benzodioxol-5-ylcarbonyl, n-3,4-methylenedioxybenzoyl piperidine, 1,3-benzodioxol-5-yl piperidin-1-yl methanone, 1-2h-1,3-benzodioxole-5-carbonyl piperidine, 1,3-bycp, tocris-1048, biomol-nt_000223, 4-20-00-01340 beilstein handbook reference
1,3-benzodioxol-5-yl(piperidin-1-yl)methanone
C13H15NO3
BXBNADAPIHHXJQ-UHFFFAOYSA-N
1370
C1CCN(CC1)C(=O)C2=CC3=C(C=C2)OCO3

Specifications

1-BCP
34023-62-6
1-bcp, 1-1,3-benzodioxol-5-ylcarbonyl piperidine, piperidine, 1-1,3-benzodioxol-5-ylcarbonyl, n-3,4-methylenedioxybenzoyl piperidine, 1,3-benzodioxol-5-yl piperidin-1-yl methanone, 1-2h-1,3-benzodioxole-5-carbonyl piperidine, 1,3-bycp, tocris-1048, biomol-nt_000223, 4-20-00-01340 beilstein handbook reference
C1CCN(CC1)C(=O)C2=CC3=C(C=C2)OCO3
233.267
233.27
50mg
C13H15NO3
BXBNADAPIHHXJQ-UHFFFAOYSA-N
1,3-benzodioxol-5-yl(piperidin-1-yl)methanone
1370
>99%
Videos