missing translation for 'onlineSavingsMsg'
Learn More

4-Azaxanthone, 97%, Thermo Scientific Chemicals

Catalog Number p-7081876
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6537-46-8
C12H7NO2
MFCD00052253
This item is not returnable. View return policy
6537-46-8
C12H7NO2
MFCD00052253

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 180°C to 182°C
Quantity 1 g
Beilstein 8657
Formula Weight 197.2
Percent Purity 97%
Chemical Name or Material 1-Azaxanthone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'