missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminocyclobutanecarboxylic acid, 95%, Thermo Scientific Chemicals
22.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
Solubility
Soluble to 100 mM in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
Solubility
Soluble to 100 mM in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 261°C (decomposition) |
Color | White |
Quantity | 1 g |
Beilstein | 2802253 |
Solubility Information | Soluble to 100 mM in water. |
Formula Weight | 115.13 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 1-Aminocyclobutanecarboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'