missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-1-naphthylamine, 98%, Acros Organics™

Brand:  Acros Organics 104650050

Code : 29

Additional Details : CAS Number : 2217-41-6 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10395730

  • £35.40 / 5g


Estimated Shipment
02-07-2018
Add to basket


Specifications

Chemical Identifiers

5,6,7,8-Tetrahydro-1-naphthylamine
2217-41-6
100.0
98%
C10H13N
1-Amino-5,6,7,8-tetrahydronaphthalene, 1-Amino-5, 6, 7
tech., 90%
97.5
Technical Grade
98%
147.22

Specifications

5,6,7,8-Tetrahydro-1-naphthylamine
2217-41-6
97.5
Glass bottle
Solubility in water: insoluble. Other solubilities: freely soluble in organic solvents (20°C)
Clear yellow to brown
275.0° to 277.0°C (713.0 mmHg)
Liquid
>110°C
98%
147.22
1-Amino-5,6,7,8-tetrahydronaphthalene, 1-Amino-5, 6, 7
tech., 90%
Authentic
100.0
Consistent with structure
1.0560g/mL
38.0°C
Technical Grade
5g
98%
C10H13N
12, 1197
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5,6,7,8-Tetrahydro-1-naphthylamine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS