missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-4-hydroxyanthraquinone 96%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Melting Point | 207.0°C to 209.0°C |
Color | Brown to Purple |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% |
Packaging | Glass Bottle |
Quantity | 5 g |
Beilstein | 14, 268 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and benzene |
Show More |
Safety and Handling
Product Identifier
- 1-Amino-4-hydroxyanthraquinone
- Danger
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Germ cell mutagenicity Category 2
- Specific target organ toxicity Category 2
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H371-May cause damage to organs.
- H304-May be fatal if swallowed and enters airways.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'