missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-(2-hydroxyethyl)piperazine, 96%, ACROS Organics™

Brand:  Acros Organics 153810010

Code : 29

Additional Details : CAS Number : 3973-70-4 Weight : 0.05100kg Transport : UN number : 3263 Chem class : 8 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10377900

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1-Amino-4-(2-hydroxyethyl)piperazine
95.0
≥95%
C6H15N3O
MFCD00006155
3973-70-4
100.0
96%
145.2
4-Amino-1-piperazineethanol

Specifications

1-Amino-4-(2-hydroxyethyl)piperazine
95.0
Authentic
Crystalline Powder and Chunks
96%
102° to 107°C
MFCD00006155
1g
3973-70-4
100.0
Glass bottle
≥95%
C6H15N3O
145.2
Yellow
4-Amino-1-piperazineethanol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Amino-4-(2-hydroxyethyl)piperazine

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS