missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Amino-4-(2-hydroxyethyl)piperazine, 96%

Brand:  Alfa Aesar™ H55134.03

UNSPSC : 12352100

Code : NEW

Additional Details : CAS Number : 3973-70-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471288

  • £109.71 / 1g


Estimated Shipment date
from Supplier 07-09-2017
Add to basket

Specifications

Chemical Identifiers

1-Amino-4-(2-hydroxyethyl)piperazine
145.2
4-Amino-1-piperazineethanol
3973-70-4
MFCD00006155

Specifications

1-Amino-4-(2-hydroxyethyl)piperazine
Powder
105° to 107°C
MFCD00006155
1g
3973-70-4
96%
145.2
White to pale brown
4-Amino-1-piperazineethanol
Documentation

Technical Documents