missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95+% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3OC6H4OCH2CH(OH)CH2NH2 |
Quantity | 1 g |
Formula Weight | 197.23 |
Percent Purity | 95% |
Chemical Name or Material | 1-Amino-3-(4-methoxyphenoxy)propan-2-ol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'