missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-54641
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22059-21-8
C4H7NO2
101.11
MFCD00009944
PAJPWUMXBYXFCZ-UHFFFAOYSA-N
1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
535
CHEBI:18053
1-aminocyclopropane-1-carboxylic acid
NC1(CC1)C(O)=O
This item is not returnable. View return policy
22059-21-8
C4H7NO2
101.11
MFCD00009944
PAJPWUMXBYXFCZ-UHFFFAOYSA-N
1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
535
CHEBI:18053
1-aminocyclopropane-1-carboxylic acid
NC1(CC1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 257°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC3H4CO2H
Quantity 250 mg
Formula Weight 101.1
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material 1-Amino-1-cyclopropanecarboxylic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'