missing translation for 'onlineSavingsMsg'
Learn More

1-Allylpiperazine, 99%, Thermo Scientific Chemicals

Catalog Number p-4089800
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13961-36-9
C7H14N2
126.2
MFCD00167970
ZWAQJGHGPPDZSF-UHFFFAOYSA-N
1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
806422
1-prop-2-enylpiperazine
C=CCN1CCNCC1
This item is not returnable. View return policy
13961-36-9
C7H14N2
126.2
MFCD00167970
ZWAQJGHGPPDZSF-UHFFFAOYSA-N
1-allylpiperazine, 1-allyl-piperazine, 1-allyl piperazine, 1-prop-2-en-1-yl piperazine, n-allyl piperazine, piperazine,1-2-propen-1-yl, prop-2-enylpiperazine, allylpiperazin, allylpiperazine, 1-allylpiperazin
806422
1-prop-2-enylpiperazine
C=CCN1CCNCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.0000g/mL
Boiling Point 181.0°C to 182.0°C
Flash Point 51.7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1
Formula Weight 126.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Allylpiperazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Allylpiperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'