missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Adamantylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Description
1-Adamantylzinc bromide is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Adamantylzinc bromide is used as pharmaceutical intermediates.
Solubility
Reacts with water.
Notes
Air Sensitive. Incompatible with oxidizing agents, bases and heat. Store under dry inert gas. Sto
1-Adamantylzinc bromide is used as pharmaceutical intermediates.
Solubility
Reacts with water.
Notes
Air Sensitive. Incompatible with oxidizing agents, bases and heat. Store under dry inert gas. Sto
Specifications
CAS | 312624-15-0 |
Density | 0.97 |
Molecular Formula | C10H15BrZn |
MDL Number | MFCD00671988 |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive; May form a precipitate |
Synonym | 1-adamantylzinc bromide, 1-adamantylzinc bromide solution, adamantan-1-yl bromo zinc, 1-adamantylzinc bromide solution, 0.5 m in thf, bromo 3r,5s,7s-adamantan-1-yl zinc, 1-adamantylzinc bromide 0.5 m in tetrahydrofuran, 1-adamantylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles |
Solubility Information | Reacts with water. |
InChI Key | PTWVHBHYYPLVCD-UHFFFAOYSA-M |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'