missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog Number 10638454
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
828-51-3
C11H16O2
180.25
MFCD00074720
JIMXXGFJRDUSRO-UHFFFAOYSA-N
1-adamantanecarboxylic acid, 1-carboxyadamantane, adamantanecarboxylic acid, adamantoic acid, 1-adamantoic acid, 3-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, tricyclo 3.3.1.1'3,7 decane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3r,5s,7s-adamantane-1-carboxylic acid
13235
adamantane-1-carboxylic acid
C1C2CC3CC1CC(C2)(C3)C(=O)O
This item is not returnable. View return policy
828-51-3
C11H16O2
180.25
MFCD00074720
JIMXXGFJRDUSRO-UHFFFAOYSA-N
1-adamantanecarboxylic acid, 1-carboxyadamantane, adamantanecarboxylic acid, adamantoic acid, 1-adamantoic acid, 3-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, tricyclo 3.3.1.1'3,7 decane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3r,5s,7s-adamantane-1-carboxylic acid
13235
adamantane-1-carboxylic acid
C1C2CC3CC1CC(C2)(C3)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 172°C to 176°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 180.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanecarboxylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Adamantanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'