missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetylpiperidine-4-carbonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 152.2 |
Chemical Name or Material | 1-Acetylpiperidine-4-carbonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'