missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-5-indolinesulfonyl chloride, 97%, Maybridge

Brand:  Maybridge MO00610DA

Code : 31

Additional Details : CAS Number : 52206-05-0 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 1a Pack group : III

Quantity 1g
Packaging Amber glass bottle

 View more versions of this product

Product Code. 10137372

  • £41.00 / 1g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Acetyl-5-indolinesulfonyl chloride
97.0
97%
259.71
52206-05-0
97%
C10H10ClNO3S
MFCD07368558

Specifications

1-Acetyl-5-indolinesulfonyl chloride
97.0
97%
C10H10ClNO3S
MFCD07368558
52206-05-0
Amber glass bottle
97%
259.71
1g
Videos
SDS