missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444440050

Code : 29

Additional Details : CAS Number : 1150561-76-4 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11976221

  • £572.90 / 5g


Estimated Shipment
19-03-2018
Add to basket


Specifications

Chemical Identifiers

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester
97%
236.08
≥96.0% (GC)
C11H17BN2O3

Specifications

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester
Crystalline Powder
97%
C11H17BN2O3
White to yellow to pale orange
1150561-76-4
≥96.0% (GC)
96% min. consistent with structure (1H-NMR)
236.08
5g
Videos
SDS