missing translation for 'onlineSavingsMsg'
Learn More

1-Acetamidoadamantane, 99+%, Thermo Scientific Chemicals

Catalog Number 10745971
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
880-52-4
C12H19NO
193.29
MFCD00074730
BCVXYGJCDZPKGV-UHFFFAOYSA-N
1-acetamidoadamantane, n-1-adamantyl acetamide, n-adamantan-1-yl acetamide, 1-acetylaminoadamantane, 1-adamantylacetamide, acetamide, n-1-adamantyl, n-adamantylacetamide, 1-acetamino adamantane, unii-5283y1voii, n-acetyl adamantamine
64153
N-(1-adamantyl)acetamide
CC(=O)NC12CC3CC(C1)CC(C3)C2
This item is not returnable. View return policy
880-52-4
C12H19NO
193.29
MFCD00074730
BCVXYGJCDZPKGV-UHFFFAOYSA-N
1-acetamidoadamantane, n-1-adamantyl acetamide, n-adamantan-1-yl acetamide, 1-acetylaminoadamantane, 1-adamantylacetamide, acetamide, n-1-adamantyl, n-adamantylacetamide, 1-acetamino adamantane, unii-5283y1voii, n-acetyl adamantamine
64153
N-(1-adamantyl)acetamide
CC(=O)NC12CC3CC(C1)CC(C3)C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 148.0°C to 150.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 12, 220
Formula Weight 193.29
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetamidoadamantane, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Acetamidoadamantane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'