missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,9-Decadiene, 97%

Brand:  Alfa Aesar™ 031207.04

Code : NEW

Additional Details : CAS Number : 1647-16-1 Weight : 0.00200kg Transport : UN number : 3295 Chem class : 1 Pack group : III

Quantity 2g

 View more versions of this product

Product Code. 11347428

  • £21.30 / 2g


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

1,9-Decadiene
138.25
UN3295
1647-16-1
MFCD00008670

Specifications

1,9-Decadiene
0.75g/mL
Liquid
169°C
MFCD00008670
2g
1647-16-1
1.4325
97%
138.25
UN3295
42° (107°F)
Videos
SDS