missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanedithiol, 99%, Thermo Scientific Chemicals

Catalog Number p-176616
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1191-62-4
C8H18S2
178.35
MFCD00003574
PGTWZHXOSWQKCY-UHFFFAOYSA-N
1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474
14493
octane-1,8-dithiol
SCCCCCCCCS
This item is not returnable. View return policy
1191-62-4
C8H18S2
178.35
MFCD00003574
PGTWZHXOSWQKCY-UHFFFAOYSA-N
1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474
14493
octane-1,8-dithiol
SCCCCCCCCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 269.0°C to 270.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HS(CH2)8SH
Refractive Index 1.5030 to 1.5050
Quantity 1 g
Specific Gravity 0.96
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 178.36
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 8-Octanedithiol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'