missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanediol, 98%, Thermo Scientific Chemicals

Catalog Number p-176608
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
629-41-4
C8H18O2
146.23
OEIJHBUUFURJLI-UHFFFAOYSA-N
69420
CHEBI:44630
octane-1,8-diol
C(CCCCO)CCCO
This item is not returnable. View return policy
629-41-4
C8H18O2
146.23
OEIJHBUUFURJLI-UHFFFAOYSA-N
69420
CHEBI:44630
octane-1,8-diol
C(CCCCO)CCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 60.0°C to 62.0°C
Color White to Yellow
Boiling Point 278.8°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula HO(CH2)8OH
Quantity 25 g
Beilstein 01, 490
Synonym 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p
Solubility Information Solubility in water: slightly soluble in water.
Viscosity 20.6 mPa.s (70°C)
Formula Weight 146.23
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1, 8-Octanediol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,8-Octanediol, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'