Learn More
Thermo Scientific Maybridge [1,8]Naphthyridine-2-carbaldehyde, 1g, 97%
Brand: Thermo Scientific Maybridge MO01212DA
[1,8]Naphthyridine-2-carbaldehyde, 1g, 97%
Code : 31
Additional Details : CAS Number : 64379-45-9 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III