missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,7-Phenanthroline

Brand:  Alfa Aesar™ 030909.06

Code : NEW

Additional Details : CAS Number : 230-46-6 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11347378

  • £224.90 / 5g


Estimated Shipment date
from Supplier 27-04-2018
Add to basket

Specifications

Chemical Identifiers

1,7-Phenanthroline
180.21
UN2811
230-46-6
MFCD00004971

Specifications

1,7-Phenanthroline
230-46-6
Crystalline
MFCD00004971
77° to 81°C
5g
Odorless
180.21
UN2811
Videos
SDS