missing translation for 'onlineSavingsMsg'
Learn More

1-(6-n-Butoxy-2-pyridyl)piperazine, Thermo Scientific™

Catalog Number p-7033785
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
902837-07-4
C13H21N3O
235.331
MFCD08061085
PPOHEYKESQFZEC-UHFFFAOYSA-N
1-6-butoxypyridin-2-yl piperazine, 1-6-n-butoxy-2-pyridyl piperazine, 1-6-butoxy pyridin-2-yl piperazine, acmc-20ap9t, 2-butoxy-6-piperazinylpyridine
28054514
1-(6-butoxypyridin-2-yl)piperazine
CCCCOC1=CC=CC(=N1)N2CCNCC2
This item is not returnable. View return policy
902837-07-4
C13H21N3O
235.331
MFCD08061085
PPOHEYKESQFZEC-UHFFFAOYSA-N
1-6-butoxypyridin-2-yl piperazine, 1-6-n-butoxy-2-pyridyl piperazine, 1-6-butoxy pyridin-2-yl piperazine, acmc-20ap9t, 2-butoxy-6-piperazinylpyridine
28054514
1-(6-butoxypyridin-2-yl)piperazine
CCCCOC1=CC=CC(=N1)N2CCNCC2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 235.33
Chemical Name or Material 1-(6-n-Butoxy-2-pyridyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'