missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadien-4-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-140064
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2883-45-6
C7H12O
112.17
MFCD00008663
UTGFOWQYZKTZTN-UHFFFAOYSA-N
1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference
17902
hepta-1,6-dien-4-ol
OC(CC=C)CC=C
This item is not returnable. View return policy
2883-45-6
C7H12O
112.17
MFCD00008663
UTGFOWQYZKTZTN-UHFFFAOYSA-N
1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference
17902
hepta-1,6-dien-4-ol
OC(CC=C)CC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 151°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2CH(OH)CH2CH=CH2
Refractive Index 1.4490 to 1.453
Quantity 1 g
Beilstein 01, 455
Specific Gravity 0.86
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 6-Heptadien-4-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,6-Heptadien-4-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'