missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diphenyl-1,3,5-hexatriene, 95%, Thermo Scientific Chemicals

Catalog Number p-122650
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1720-32-7
C18H16
232.32
MFCD00004793
BOBLSBAZCVBABY-WPWUJOAOSA-N
1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
5376733
CHEBI:51594
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
This item is not returnable. View return policy
1720-32-7
C18H16
232.32
MFCD00004793
BOBLSBAZCVBABY-WPWUJOAOSA-N
1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
5376733
CHEBI:51594
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 199°C to 203°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCH=CHCH=CHC6H5
Quantity 1 g
Beilstein 05, 691
Formula Weight 232.32
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1,6-Diphenyl-1,3,5-hexatriene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'