missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromo-2-naphthol, 99%, Thermo Scientific™

Catalog Number p-102555
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
16239-18-2
C10H6Br2O
301.97
MFCD00003870
VKESFYLPKHQOOA-UHFFFAOYSA-N
1,6-dibromo-2-naphthol, 2-naphthalenol, 1,6-dibromo, 2-naphthol, 1,6-dibromo, 1,6-dibromo-2-hydroxynaphthalene, 1,6-dibromo-beta-naphthol, 1,6-dibromo-.beta.-naphthol, pubchem22920, 2-naphthol,6-dibromo, 1,6 dibromo-2-napthol, acmc-1c8jh
27768
1,6-dibromonaphthalen-2-ol
OC1=CC=C2C=C(Br)C=CC2=C1Br
This item is not returnable. View return policy
16239-18-2
C10H6Br2O
301.97
MFCD00003870
VKESFYLPKHQOOA-UHFFFAOYSA-N
1,6-dibromo-2-naphthol, 2-naphthalenol, 1,6-dibromo, 2-naphthol, 1,6-dibromo, 1,6-dibromo-2-hydroxynaphthalene, 1,6-dibromo-beta-naphthol, 1,6-dibromo-.beta.-naphthol, pubchem22920, 2-naphthol,6-dibromo, 1,6 dibromo-2-napthol, acmc-1c8jh
27768
1,6-dibromonaphthalen-2-ol
OC1=CC=C2C=C(Br)C=CC2=C1Br
CAS Min % 98.5
CAS Max % 100.0
Melting Point 105.0°C to 107.0°C
Color Brown or Pink to Purple
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C10H5OH
Quantity 25 g
Beilstein 06, 652
Formula Weight 301.96
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 6-Dibromo-2-naphthol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,6-Dibromo-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'