missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 98+%, Thermo Scientific™

Catalog Number p-60362
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
13242-55-2
C12H16O8
288.25
MFCD00063247
BAKQMOSGYGQJOJ-UHFFFAOYNA-N
triacetyllevoglucosan, 1,6-anhydro-, a-d-glucopyranose 2,3,4-triacetate, 2,3,4-tri-o-acetyl-1,6-anhydro-, a-d-glucopyranose, 2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate
134159065
[(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate
CC(=O)OC1C2COC(O2)C(OC(C)=O)C1OC(C)=O
This item is not returnable. View return policy
13242-55-2
C12H16O8
288.25
MFCD00063247
BAKQMOSGYGQJOJ-UHFFFAOYNA-N
triacetyllevoglucosan, 1,6-anhydro-, a-d-glucopyranose 2,3,4-triacetate, 2,3,4-tri-o-acetyl-1,6-anhydro-, a-d-glucopyranose, 2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate
134159065
[(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate
CC(=O)OC1C2COC(O2)C(OC(C)=O)C1OC(C)=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 111.0°C to 112.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation − 66.00
Solubility Information Solubility in water: soluble.
Formula Weight 288.26
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'