missing translation for 'onlineSavingsMsg'
Learn More

1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™

Catalog Number p-4391497
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
950603-35-7
C9H10F3N3O2S
281.25
MFCD11101440
PWQMNLMMMCREOV-UHFFFAOYSA-N
1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid
33589561
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F
This item is not returnable. View return policy
950603-35-7
C9H10F3N3O2S
281.25
MFCD11101440
PWQMNLMMMCREOV-UHFFFAOYSA-N
1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid
33589561
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 281.25
Percent Purity 97%
Chemical Name or Material 1-[5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-yl]piperidine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'