missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl-1H-pyrazole-3-carbaldehyde, ≥97%, Maybridge

Brand:  Maybridge CC12604DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 25016-10-8 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10770704

  • £73.00 / 1g


Estimated Shipment date
from Supplier 04-07-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

1, 5-Dimethyl-1H-pyrazole-3-carbaldehyde
96.5
97%
C6H8N2O
MFCD04967221
25016-10-8
100.0
97%
124.14

Specifications

124.14
100.0
25016-10-8
C6H8N2O
Amber Glass Bottle
MFCD04967221
1g
1, 5-Dimethyl-1H-pyrazole-3-carbaldehyde
97%
96.5
97%
Documentation

Technical Documents