missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Dimethyl-1H-pyrazol-3-yl)methylamine, 97%, Maybridge

Brand:  Maybridge CC12613DE

Code : 31

Additional Details : CAS Number : 423768-52-9 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 10610286

  • £639.00 / 5g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

(1, 5-Dimethyl-1H-pyrazol-3-yl)methylamine
96.5
97%
C6H11N3
MFCD03422531
423768-52-9
100.0
97%
125.17

Specifications

(1, 5-Dimethyl-1H-pyrazol-3-yl)methylamine
423768-52-9
100.0
97%
125.17
5g
96.5
97%
C6H11N3
MFCD03422531
Videos
SDS