missing translation for 'onlineSavingsMsg'
Learn More

1,5-Cyclooctadiene, 99+%, stabilized, purified by redistillation, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 111418000

Code : 29

Additional Details : CAS Number : 111-78-4 Weight : 0.75400kg Transport : UN number : 2520 Chem class : 3 Pack group : III

Quantity 800mL
Packaging AcroSeal™ Glass Bottle

 View more versions of this product

Product Code. 11944431

  • £196.20 / 800mL


Estimated Shipment
22-06-2018
Add to basket

Specifications

Chemical Identifiers

1, 5-Cyclooctadiene
99.0
≥99%
C8H12
COD
111-78-4
100.0
99+%
108.18

Specifications

1, 5-Cyclooctadiene
38°C
Clear colorless
800mL
99.0
Authentic
1.4920 to 1.4940
Liquid
99+%
108.18
0.8800g/mL
151.0°C (101.3 kPa)
− 69.5°C
111-78-4
100.0
AcroSeal™ Glass Bottle
100 to 200ppm octadecyl 3-(3«, 5«-di-tert-butyl-4«-hydroxyphenyl)propionate
≥99%
C8H12
COD
Videos
SDS