missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(5-Bromo-2,3-dihydro-1H-indol-1-yl)ethan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 240.1 |
Percent Purity | 97% |
Chemical Name or Material | 1-(5-Bromo-2, 3-dihydro-1H-indol-1-yl)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'