missing translation for 'onlineSavingsMsg'
Learn More

1,5-Bis(methylamino)-3-oxapentane, 98%, Thermo Scientific Chemicals

Catalog Number p-4510393
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2620-27-1
C6H16N2O
132.21
VXPJBVRYAHYMNY-UHFFFAOYSA-N
2724770
N-methyl-2-[2-(methylamino)ethoxy]ethanamine
CNCCOCCNC
This item is not returnable. View return policy
2620-27-1
C6H16N2O
132.21
VXPJBVRYAHYMNY-UHFFFAOYSA-N
2724770
N-methyl-2-[2-(methylamino)ethoxy]ethanamine
CNCCOCCNC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.8720g/mL
Boiling Point 76.0°C to 78.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3NHCH2CH2OCH2CH2NHCH3
Refractive Index 1.4370 to 1.4390
Quantity 1 g
Specific Gravity 0.872
Synonym 2,2'-oxybis n-methylethanamine, ethanamine,2,2'-oxybis n-methyl, ethanamine, 2,2'-oxybis n-methyl, 1,5-bis methylamino-3-oxapentane, methyl 2-2-methylamino ethoxy ethyl amine, bmaop, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 132.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 5-Bis(methylamino)-3-oxapentane
Show More Show Less
compliance-icons
Product Identifier
  • 1,5-Bis(methylamino)-3-oxapentane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'