missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane, 95%, Thermo Scientific™

Catalog Number p-2695413
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
238403-48-0
C10H13F3N4
246.24
MFCD00728997
XVAZHAVSZFQOOQ-UHFFFAOYSA-N
1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazepane, 1-4-trifluoromethyl-2-pyrimidinyl-1,4-diazepane, 1-4-trifluoromethyl pyrimid-2-yl homopiperazine, 1-4-trifluoromethyl pyrimidin-2-yl homopiperazine, 1h-1,4-diazepine,hexahydro-1-4-trifluoromethyl-2-pyrimidinyl, maybridge1_000269, 1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazaperhydroepine
2782067
1-[4-(trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane
FC(F)(F)C1=NC(=NC=C1)N1CCCNCC1
This item is not returnable. View return policy
238403-48-0
C10H13F3N4
246.24
MFCD00728997
XVAZHAVSZFQOOQ-UHFFFAOYSA-N
1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazepane, 1-4-trifluoromethyl-2-pyrimidinyl-1,4-diazepane, 1-4-trifluoromethyl pyrimid-2-yl homopiperazine, 1-4-trifluoromethyl pyrimidin-2-yl homopiperazine, 1h-1,4-diazepine,hexahydro-1-4-trifluoromethyl-2-pyrimidinyl, maybridge1_000269, 1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazaperhydroepine
2782067
1-[4-(trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane
FC(F)(F)C1=NC(=NC=C1)N1CCCNCC1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 246.23
Percent Purity 95%
Chemical Name or Material 1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1, 4-diazepane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'