missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-(Trifluoromethoxy)phenyl]-3-[3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64037.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458656

  • £460.04 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-(Trifluoromethoxy)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
364.24
MFCD04280569

Specifications

1-[4-(Trifluoromethoxy)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
Solid
364.24
5g
97%
MFCD04280569
Videos
SDS