missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-(Trifluoromethoxy)phenyl]-3-(2,3,4-trifluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H64797.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419156

  • £79.71 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-(Trifluoromethoxy)phenyl]-3-(2,3,4-trifluorophenyl)urea
350.22
MFCD26960710

Specifications

1-[4-(Trifluoromethoxy)phenyl]-3-(2,3,4-trifluorophenyl)urea
Solid
350.22
1g
97%
MFCD26960710
Videos
SDS