missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-[4-(Trifluoromethoxy)phenyl]-3-(2,3,4-trifluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H64797.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429156

  • £398.56 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

1-[4-(Trifluoromethoxy)phenyl]-3-(2,3,4-trifluorophenyl)urea
350.22
MFCD26960710

Specifications

1-[4-(Trifluoromethoxy)phenyl]-3-(2,3,4-trifluorophenyl)urea
Solid
350.22
5g
97%
MFCD26960710
Videos
SDS