missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-tert-Butylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H65129.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449396

  • £79.71 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-tert-Butylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
370.8
MFCD27975722

Specifications

1-(4-tert-Butylphenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
97%
MFCD27975722
Solid
370.8
1g
Videos
SDS