missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Pyridyl)pyridinium chloride hydrochloride, tech. 85%

Brand:  Alfa Aesar™ H55351.06

Code : NEW

Additional Details : CAS Number : 5421-92-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15422056

  • £15.90 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Pyridyl)pyridinium chloride hydrochloride
Technical Grade
MFCD00012821
5421-92-1
229.11
3786515

Specifications

1-(4-Pyridyl)pyridinium chloride hydrochloride
Yellow to brown
5g
Technical Grade
85%
MFCD00012821
Hygroscopic
166° to 170°C
5421-92-1
Solid
229.11
3786515
Videos
SDS